O-Ring seals for flat flanges
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06248010 06248015 06248215 06248210 06248206 06248006 06248020 06248220
201-0152EA 15.3 EUR
201-0152 LENZ06248015 LENZ06248215 LENZ06248210 LENZ06248206 201-0151 LENZ06248020 LENZ06248220
O-Ring seals for flat flanges
Afdichtingen Afdichtringen
Silicone or Silicone PTFE-coated.
  • Excellent chemical resistance
  • Excellent temperature resistance
Seals for flat flanges LF 60-200.