Stirring shafts
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06300510 06300516
LENZ06300510EA 33 EUR
LENZ06300510 LENZ06300516
Stirring shafts
Roerders Roerassen
Stirring shafts made of precision calibrated glass tube.
  • Ground and polished bearing surface
  • Interchangeable
  • Made of DURAN® tubing
Order Now
Specificaties
Environmentally Preferable