Slangen, Versilon® 2001
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AE300003 AE300007 AE300012 AE300017 AE300022 AE300027 AE300038 AE300046 AE300053 AE300064
228-1268EA 178 EUR
228-1268 228-1269 228-1270 228-1271 228-1272 228-1273 228-1274 228-1275 228-1276 228-1277
Slangen, Versilon® 2001
Slangen Transfer Tubing
Versilon®, clear, for peristaltic pumps.
  • Plasticiser and oil free, does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Clear for easy visual flow monitoring
Versilon® 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer/oil-free tubing product. It is resistant to a wide range of fluids that typically destroy PVC products. Its chemical resistance allows it to be used in applications with aggressive chemicals such as polar solvents, benefiting the user with less downtime and tubing changes. It is ideal for a broad range of demanding applications including peristaltic pump systems, soap and detergents dispensing, ink transfer, water purification lines, food and beverage, and chemical transfer.
Temperature resistance: −78 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88
Certificaten: Complies with FDA 21 CFR, 177.2600 and REACH