VWR®, Mediumflessen met dop
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
391-0283EA 158 EUR
391-0283 391-0284 391-0286 391-0299
VWR®, Mediumflessen met dop
Flessen Mediumflessen
PS, PC of PETG, met PP-schroefdop, steriel.
  • Sloped design improves bottle stability when placed on a work surface
  • Narrow bottle shoulder, combined with moulded finger grips, improves the ability to securely grip the bottle with gloved hands
  • Easy Grasp bottle cap facilitates uncapping, tightening and bottle stacking, as well as providing a convenient gripping surface while transporting
  • Polystyrene (PS) bottles are supplied with a white cap
  • Polycarbonate (PC) bottles are supplied with a blue cap
  • PETG bottles are supplied with a green cap
VWR® Collection-More Product Information