Pasteur pipettes
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9250101 9250201 9260101 9260201
HIRS9250101EA 65.1 EUR
HIRS9250101 HIRS9250201 HIRS9260101 HIRS9260201
Pasteur pipettes
Pipetten Pasteur-/transferpipetten
Glass pasteur pipettes with fine drawn tips.
  • With or without cotton plug
  • Short or long form
Certificaten: According to ISO 7712.