Very high pressure (VHP) fittings, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHP-200 VHP-205 VHP-320X VHP-325 VHP-320 VHP-305 VHP-325X VHP-3200
554-1821EA 46.6 EUR
554-1821 554-1823 554-5696 554-1997 554-1996 554-1995 554-5697 554-2270
Very high pressure (VHP) fittings, Upchurch Scientific®
Fittingen en connectoren
Reusable fittings made from stainless steel and proprietary PEEK polymer blend. Reusable up to 10 cycles without tubing damage or impact on pressure holding ability or carry-over (5 cycles if tightened to 1,6 N.m). Pressure rating up to 30 000 psi (2070 bar). Required torque 10 in-lbs (1,10 N-m)/14 in-lbs (1,6 N-m).
Stainless steel fittings have a double compression ferrule design. Pressure rating up to 30 000 psi (2070 bar). Required torque 20 in-lbs (2,25 N-m).