Kolven, vlakke bodem, wijde hals, met rolrand
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07050037 07050049 07050058 07050070 07050073 07050074
LENZ07050037EA 144 EUR
LENZ07050037 LENZ07050049 LENZ07050058 LENZ07050070 LENZ07050073 LENZ07050074
Kolven, vlakke bodem, wijde hals, met rolrand
Kolven Platbodemkolven
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
Certificaten: Acc. to DIN ISO 24450