Syringes, SampleLock™
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
81656 81056 86326 86336 86346 81256 81356 81456 81556 80956 81156 CAL81156
HAMI81656EA 387 EUR
HAMI81656 HAMI81056 HAMI86326 HAMI86336 HAMI86346 HAMI81256 HAMI81356 HAMI81456 HAMI81556 HAMI80956 HAMI81156 HAMICAL81156
Syringes, SampleLock™
Spuiten Injectiespuiten voor chromatografie
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.