Safety pipette fillers, standard / universal / flip
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9910101 9910103 9910105
HIRS9910101EA 17.2 EUR
HIRS9910101 HIRS9910103 HIRS9910105
Safety pipette fillers, standard / universal / flip
Pipetten Pipetvullerballonnen
Safety pipette fillers made of natural rubber.
  • Suitable for graduated and volumetric pipettes