Filterbuizen, Allihn
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07220001 07220002 07220003 07220004
LENZ07220001EA 33.4 EUR
LENZ07220001 LENZ07220002 LENZ07220003 LENZ07220004
Filterbuizen, Allihn
Trechters Filtertrechters Microfiltertrechters
Made of DURAN®, borosilicate glass 3,3.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.