Sleeves for filter crucibles
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
462026 462231
BRND462026EA 51 EUR
BRND462026 BRND462231
Sleeves for filter crucibles
Steunen Steunhoezen
Sleeves made of rubber (EPDM) suitable for filter crucibles.