Flessen, smalle hals, vierkant, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312018-1000 312018-0125 312018-0250 312018-0500
NALG312018-1000EA 0 EUR
NALG312018-1000 NALG312018-0125 NALG312018-0250 NALG312018-0500
Flessen, smalle hals, vierkant, met schroefdop, Nalgene®
Flessen Kunststof flessen
HDPE, translucent, with PP screw cap.
  • Good chemical resistance
  • Good temperature resistance
  • Leakproof
  • Non sterile
  • Not autoclavable
Ideal for storing and shipping liquids.
These translucent bottles provide visible liquid levels and molded-in graduations for easy measuring. Square narrow mouth bottles with polypropylene closures feature space-saving designs and drip-proof necks for convenient dispensing.
Verpakking: Bottles and closures are bulk-packed in separate polybags within the same carton.