Verpakkingsflessen, smalle hals, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312087-0001 312087-0002 312087-0004 312087-0008 312087-0016 312087-0032
NALG312087-0001EA 1000 EUR
NALG312087-0001 NALG312087-0002 NALG312087-0004 NALG312087-0008 NALG312087-0016 NALG312087-0032
Verpakkingsflessen, smalle hals, met schroefdop, Nalgene®
Flessen Kunststof flessen
PPCO, translucent, with PP screw cap.
  • Excellent clarity
  • Impact and chemical resistance
  • Leakproof
  • Autoclavable
These bottles have a robust construction for packaging use.
Certificaten: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpakking: Bottles and closures are packed in separate polybags within the same carton.
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement IRECS; ISO 14001 (Certificate Registration No. EMS 752603)