Flessen, smalle hals, rond, PP, IP2, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312099-0001 312099-0004 312099-0008 312099-0016 312099-0032
NALG312099-0001EA 1740 EUR
NALG312099-0001 NALG312099-0004 NALG312099-0008 215-4637 NALG312099-0032
Flessen, smalle hals, rond, PP, IP2, met schroefdop, Nalgene®
Flessen Kunststof flessen
HDPE, translucent with PP screw cap
  • Good chemical resistance
  • Screw cap supplied assembled on the bottle
  • Leakproof
Bottles with an average of less than 30 particles per ml at 0,3 µm and greater. Excellent for storing high-purity chemicals. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with Nalgene® specifications. Low particulate IP2 bottles can be used in compliance with the regulations for customers who are designing, assembling and certifying their own combination packaging.