Flessen, brede hals, LDPE, rond, met schroefdop, laboratoriumkwaliteit, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312103-0004 312103-0016
NALG312103-0004EA 877 EUR
NALG312103-0004 NALG312103-0016
Flessen, brede hals, LDPE, rond, met schroefdop, laboratoriumkwaliteit, Nalgene®
Flessen Kunststof flessen
LDPE, translucent with PP screw cap.
  • Good chemical resistance, heat resistant up to +80 °C
  • Leakproof closure, so they are suitable for use with liquids
  • Flexible, excellent impact resistance
Can be used anywhere in the laboratory and for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.