Ethylammonium chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Aminoethane hydrochloride, Ethylamine hydrochloride
214255000 214250025
ACRO214255000EA 56.9 EUR
ACRO214255000 ACRO214250025
Ethylammonium chloride 99%
Ethylammonium chloride
Formule: C₂H₈ClN
MW: 81.54 g/mol
Smeltpunt: 105…108 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012885
CAS nummer: 557-66-4
EINECS: 209-182-8
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 %