2-Amino-1,3,5-triazine 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
295240010 295240050 AC29524.1 AC29524.10
ACRO295240010EA 52.1 EUR
ACRO295240010 ACRO295240050 MAYBAC29524.1 MAYBAC29524.10
2-Amino-1,3,5-triazine 98%
2-Amino-1,3,5-triazine
Formule: C₃H₄N₄
Smeltpunt: 219…224 °C
MDL Nummer: MFCD00023179
CAS nummer: 4122-04-7
EINECS: 223-920-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %