2,4-Toluenediisocyanate ≥80% (remainder 2,6-diisocyanate), tech.
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 80%
CAS No.: 584-84-9
Molecular Formula: C9H6N2O2
Formula Weight: 174.16
Flash Point: 132° (270°F)
Physical Form: Liquid
Density: 1.225
Refractive Index: 1.567
Sensitivity: Moisture Sensitive
Appearance: Colourless
Melting Point: 19.5-21.5°
Boiling Point: 251°
Beilstein Reference: 744602
Merck Reference: 14,9530
UN No.: 2078
MDL No.: MFCD00002011
Formule:
C₉H₆N₂O₂ Kookpunt: 251 °C Dichtheid: 1,225 Vlampunt: 132 °C (270 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002011 CAS nummer: 584-84-9 EINECS: 209-544-5 UN: 2078 ADR: 6.1,II Merck Index: 14,9530 |