Rose's metal (Bi:Pb:Sn 50:25:25 wt%), ingot
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
40516.22
40516.36
40516.22EA
169
EUR
40516.22
40516.36
Rose's metal (Bi:Pb:Sn 50:25:25 wt%), ingot
Rose's metal
Molecular Formula: Bi:Pb:Sn; 50:25:25 wt%
Physical Form: Low melting alloy
Melting Point: 95-96°
MDL No.: MFCD00804027