3,4-Dimethylphenylhydrazinium chloride ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
(3,4-Dimethylphenyl)hydrazine hydrochloride, 3,4-Dimethylphenylhydrazine hydrochloride, 1-(3,4-Dimethylphenyl)hydrazine hydrochloride
A13027.09
A13027.18
A13027.30
A13027.09EA
42.2
EUR
A13027.09
A13027.18
A13027.30
3,4-Dimethylphenylhydrazinium chloride ≥98%
3,4-Dimethylphenylhydrazinium chloride
Purity: 98%
CAS No.: 60481-51-8
Molecular Formula: C8H12N2.HCl
Formula Weight: 172.66
Melting Point: ca 195° dec.
Beilstein Reference: 6096060
Formule:
C₈H₁₃ClN₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00052270 CAS nummer: 60481-51-8 |