Colecalciferol 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Vitamin D3, Colecalciferol
B22524.06
B22524.03
B22524.06EA
146
EUR
B22524.06
B22524.03
Colecalciferol 99%
Colecalciferol
Purity: 99%
CAS No.: 67-97-0
Molecular Formula: C27H44O
Formula Weight: 384.65
Storage Temperature: 2°C to 8°C
Melting Point: 83-87°
Beilstein Reference: 2339331
Merck Reference: 14,10019
Optical Rotation: +108° (c=0.8 in ethanol)
UN No.: 2811
MDL No.: MFCD00078131
Formule:
C₂₇H₄₄O Smeltpunt: 83…87 °C |
MDL Nummer:
MFCD00078131 CAS nummer: 67-97-0 EINECS: 200-673-2 UN: 2811 ADR: 6.1,II Merck Index: 14,10019 |