Ethyl thiooxamate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate

H31447.03 H31447.06 H31447.14
H31447.03EA 36.7 EUR
H31447.03 H31447.06 H31447.14
Ethyl thiooxamate 95%
Ethyl thiooxamate

Purity: 95% CAS No.: 16982-21-1 Molecular Formula: C4H7NO2S Formula Weight: 133.17 Melting Point: 61-65° MDL No.: MFCD00074903

Formule: C₄H₇NO₂S
MW: 133.17 g/mol
Smeltpunt: 61…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074903
CAS nummer: 16982-21-1
EINECS: 605-564-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR