4-(Trifluoromethyl)phenylhydrazine 95%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 95%
CAS No.: 368-90-1
Molecular Formula: C7H7F3N2
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Melting Point: 60-64°
Boiling Point: 119-121°/17mm
Beilstein Reference: 743759
MDL No.: MFCD00042508
Formule:
C₇H₇F₃N₂ Kookpunt: 119…121 °C (17 mmHg) Smeltpunt: 60…64 °C |
MDL Nummer:
MFCD00042508 CAS nummer: 368-90-1 |