(1S,2S)-(-)-1,2-Diphenylethylenediamine 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
(1S,2S)-(-)-1,2-Diphenylethylenediamine, (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, (S,S)-DPEN
L12968.03
L12968.MD
L12968.03EA
58.3
EUR
L12968.03
L12968.MD
(1S,2S)-(-)-1,2-Diphenylethylenediamine 97%
(1S,2S)-(-)-1,2-Diphenylethylenediamine
Purity: 97%
CAS No.: 29841-69-8
Molecular Formula: C14H16N2
Formula Weight: 212.30
Storage Temperature: 2°C to 8°C
Melting Point: 80-84°
Beilstein Reference: 3201645
Optical Rotation: -102° (c=1 in methanol)
UN No.: 3259
MDL No.: MFCD00082751
Formule:
C₁₄H₁₆N₂ MW: 212.29 g/mol Smeltpunt: 80…84 °C |
MDL Nummer:
MFCD00082751 CAS nummer: 29841-69-8 ADR: 8 III |