4-Iodophenylhydrazine 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (4-Iodophenyl)hydrazine

L14806.03 L14806.06
L14806.03EA 89.1 EUR
L14806.03 L14806.06
4-Iodophenylhydrazine 95%
4-Iodophenylhydrazine

Purity: 95% CAS No.: 13116-27-3 Molecular Formula: C6H7IN2 Formula Weight: 234.04 Storage Temperature: 2°C to 8°C Melting Point: 99-103° Beilstein Reference: 742475 MDL No.: MFCD00052688

Formule: C₆H₇IN₂
MW: 234.04 g/mol
Smeltpunt: 99…103 °C
MDL Nummer: MFCD00052688
CAS nummer: 13116-27-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR