o-Tolylhydrazinium chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: o-Tolylhydrazine hydrochloride, 1-(2-Methylphenyl)hydrazine hydrochloride, 2-Tolylhydrazine hydrochloride, 1-(2-Tolyl)hydrazine hydrochloride, 2-Methylphenylhydrazinium chloride, 2-Methylphenylhydrazine hydrochloride
BTB07667.1 BTB07667.50
MAYBBTB07667.1EA 0 EUR
MAYBBTB07667.1 MAYBBTB07667.50
o-Tolylhydrazinium chloride
o-Tolylhydrazinium chloride
Formule: C₇H₁₁ClN₂
MW: 158.63 g/mol
MDL Nummer: MFCD00012925
CAS nummer: 635-26-7