2,4-Dimethylphenylhydrazinium chloride
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB08675.1
BTB08675.10
BTB08675.50
MAYBBTB08675.1EA
0
EUR
MAYBBTB08675.1
MAYBBTB08675.10
MAYBBTB08675.50
2,4-Dimethylphenylhydrazinium chloride
2,4-Dimethylphenylhydrazinium chloride
Formule:
C₈H₁₃ClN₂ |
MDL Nummer:
MFCD00013381 CAS nummer: 60480-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...