2,3-Dimethylphenylhydrazinium chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB08677.1 BTB08677.50
MAYBBTB08677.1EA 0 EUR
MAYBBTB08677.1 MAYBBTB08677.50
2,3-Dimethylphenylhydrazinium chloride
2,3-Dimethylphenylhydrazinium hydrochloride
Formule: C₈H₁₃ClN₂
MDL Nummer: MFCD00013380
CAS nummer: 56737-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR