4-Iodophenylhydrazine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(4-Iodophenyl)hydrazine
RF01374DA
RF01374EA
MAYBRF01374DAEA
0
EUR
MAYBRF01374DA
MAYBRF01374EA
4-Iodophenylhydrazine
4-Iodophenylhydrazine
Formule:
C₆H₇IN₂ |
MDL Nummer:
MFCD00052688 CAS nummer: 13116-27-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...