4-Chloro-2-cyclohexylphenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB00775.1 SB00775.10 SB00775.50
MAYBSB00775.1EA 0 EUR
MAYBSB00775.1 MAYBSB00775.10 MAYBSB00775.50
4-Chloro-2-cyclohexylphenol
4-Chloro-2-cyclohexylphenol
Formule: C₁₂H₁₅ClO
CAS nummer: 13081-17-9
EINECS: 235-988-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR