Citroenzuur 50%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
4.80855.1000 4.80855.9950 4.80855.9025 4.80855.9180
4.80855.1000EA 114 EUR
4.80855.1000 4.80855.9950 4.80855.9025 4.80855.9180
Citroenzuur 50%, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Citroenzuur
Formule: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Kookpunt: 310
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011669
CAS nummer: 77-92-9
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM