4-(4-Nitrobenzyl)pyridine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

30548.01 30548.02
SERA30548.01EA 0 EUR
SERA30548.01 SERA30548.02
4-(4-Nitrobenzyl)pyridine
4-(4-Nitrobenzyl)pyridine
Formule: C₁₂H₁₀N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006445
CAS nummer: 1083-48-3
EINECS: 214-108-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR