1,3-Dibromo-5-fluoro-2-iodobenzene
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
TL00652.1 TL00652.10 TL00652.50
MAYBTL00652.1EA 0 EUR
MAYBTL00652.1 MAYBTL00652.10 MAYBTL00652.50
1,3-Dibromo-5-fluoro-2-iodobenzene
1,3-Dibromo-5-fluoro-2-iodobenzene
Formule: C₆H₂Br₂FI
Kookpunt: 286 °C (1013 hPa)
Smeltpunt: 135…135,6 °C
MDL Nummer: MFCD00042232
CAS nummer: 62720-29-0
Order Now
Specificaties
Environmentally Preferable