Fluoranthene

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

48535.EA 56.6 EUR
48535.
Fluoranthene
Fluoranthene
Formule: C₁₆H₁₀
MW: 202,26 g/mol
MDL Nummer: MFCD00001184
CAS nummer: 206-44-0
EINECS: 205-912-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR