2,5-Dibromoaniline 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
155030050 155030250
ACRO155030050EA 42.8 EUR
ACRO155030050 ACRO155030250
2,5-Dibromoaniline 97%
2,5-Dibromoaniline
Formule: C₆H₅Br₂N
Kookpunt: 281 °C (1013 hPa)
Smeltpunt: 51…56 °C
Vlampunt: 109 °C
MDL Nummer: MFCD00007636
CAS nummer: 3638-73-1
EINECS: 222-865-5
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to dark grey Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure