Conische centrifugebuizen, conische bodem, met rand
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1387 1388
212-0208EA 2.9 EUR
212-0208 KART1388
Conische centrifugebuizen, conische bodem, met rand
Buisjes Centrifugebuisjes
PMP (TPX®), crystal clear.
  • Temperature resistance up to +170 °C
  • Autoclavable
  • High chemical resistance
  • Graduated
Conical bottom with rim.