Knofler-Bohm-extractors
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05375025 05375043 05375049 05375058 05410125 05410149 05410158
LENZ05375025EA 294 EUR
LENZ05375025 LENZ05375043 LENZ05375049 LENZ05375058 LENZ05410125 LENZ05410149 LENZ05410158
Knofler-Bohm-extractors
Extractors Soxhlet-extractors en Soxhlet-type extractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
The condensate passes the extraction sample and flows back into the flask continuously without interrupting the extraction process, i.e. the solvent is recirculated continuously.