Microcentrifugebuisjes, met schroefdoppen, Molecular BioProducts
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3468 3461 3463 3431-11 3460A 3462 3464 3466 3431A 3460 3461A 3463A 3464A 3468A 3470
MLBP3468EA 141 EUR
MLBP3468 525-1500 525-1501 525-1511 MLBP3460A MLBP3462 MLBP3464 MLBP3466 MLBP3431A MLBP3460 MLBP3461A MLBP3463A MLBP3464A MLBP3468A MLBP3470
Microcentrifugebuisjes, met schroefdoppen, Molecular BioProducts
Buisjes Microcentrifugebuisjes
Polypropylene, natural or amber.
  • O-ring in the screw cap helps prevent leaks under demanding conditions
  • Very good chemical resistance
  • Very good temperature resistance
Micro tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes.