Doseerflessen, smalle hals, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
247015409 247016302
SCOT247015409EA 51.8 EUR
SCOT247015409 SCOT247016302
Doseerflessen, smalle hals, DURAN®
Flessen Doseerflessen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High thermal resistance, autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Bottles, narrow mouth, ungrounded neck, with bottom tubulature.
With printed retrace code (batch identification).