Flessen, smalle hals, rond, HDPE, Boston-type, met schroefdraad, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
362002-0002 362002-0004 362002-0001 362002-0008 362002-0016
NALG362002-0002EA 874 EUR
NALG362002-0002 NALG362002-0004 NALG362002-0001 NALG362002-0008 NALG362002-0016
Flessen, smalle hals, rond, HDPE, Boston-type, met schroefdraad, Nalgene®
Flessen Kunststof flessen
HDPE, doorschijnend met PP-schroefdop.
  • Goede chemische bestendigheid
  • Temperatuurbestendig van −100 tot +120 °C
  • Lekvrij
Ideal for storing, packaging and transporting liquids. Robust, strong and stable.
Certificaten: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.