Flessen, brede hals, HDPE, economy, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
332189-0004 332189-0008 332189-0016 332189-0032 332189-0002
331-4261EA 452 EUR
331-4261 331-4262 331-4263 331-4264 331-0060
Flessen, brede hals, HDPE, economy, met schroefdop, Nalgene®
Flessen Kunststof flessen
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Economical, break resistant disposable bottles for safe sample collection, transport and storage. Suitable for long-term storage in the absence of direct UV radiation.