Distilling receivers, Bredt, bent
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05280001 05280004
271-2521EA 210 EUR
271-2521 271-2522
Distilling receivers, Bredt, bent
Onderdelen voor destillatie Ontvangers voor destillatie
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With four flasks NS 14/23, with bent adapter (105°) vacuum connection: GL 18.