Distilling receivers, Anschütz-Thiele, straight
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05295001 05295004 05295104 05295014 05295101 05295114
271-2511EA 247 EUR
271-2511 271-2512 LENZ05295104 LENZ05295014 LENZ05295101 LENZ05295114
Distilling receivers, Anschütz-Thiele, straight
Onderdelen voor destillatie Ontvangers voor destillatie
Made of DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receivers with straight adapter, graduated, with NS socket and NS cone, vacuum connection thread GL 18.