Distilling receivers, Anschütz-Thiele, bent
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05298001 05298004 05298014 05298101 05298104 05298114
271-2516EA 262 EUR
271-2516 271-2517 LENZ05298014 LENZ05298101 LENZ05298104 LENZ05298114
Distilling receivers, Anschütz-Thiele, bent
Onderdelen voor destillatie Ontvangers voor destillatie
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receivers with bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18, NS stopcock with solid glass plug.