Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
212276807 212277306 212278602
201-1902EA 106 EUR
201-1902 214-0237 214-0238
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
Kolven Erlenmeyers
DURAN®, borosilicaatglas 3,3, helder.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Met gerolde rand en schrijfveld. Zonder schaalverdeling.