Precisiethermometers voor distillatie
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05065006 05065001 05065004 05065005 05065002 05065003
LENZ05065006EA 81.6 EUR
LENZ05065006 LENZ05065001 LENZ05065004 LENZ05065005 LENZ05065002 LENZ05065003
Precisiethermometers voor distillatie
Thermometers Analoge thermometers
Precision distillation thermometers with cone NS 14/23, a top end which was melted round, frosted glass scale with black scale.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.