Bakjes, met schouder en deksel, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
242073408 242074507 242075109
SCOT242073408EA 37.6 EUR
SCOT242073408 216-0284 216-3118
Bakjes, met schouder en deksel, DURAN®
Potjes Potjes voor algemeen gebruik
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Jar can allows the storage and transport of materials. The matching lid prevents turbulence during transport or contamination.
Certificaten: According to DIN 12340.