Screw caps, for DIN 168 bottles, plastilab®
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15950 15990 16030 16250 16290
KART15950EA 174 EUR
KART15950 KART15990 KART16030 KART16250 KART16290
Screw caps, for DIN 168 bottles, plastilab®
Sluitingen Flesdoppen
Screw caps made of PP, it has cap tag to avoid sample contamination in transit.
Standard screw caps for polyethylene bottles manufactured to DIN 168.