Stolpen, met genormaliseerd slijpstuk, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
244655907 244656106 244656903
SCOT244655907EA 250 EUR
SCOT244655907 SCOT244656106 SCOT244656903
Stolpen, met genormaliseerd slijpstuk, DURAN®
Potjes Stolpen
DURAN®, borosilicate glass 3.3, clear.
  • Good chemical and physical properties
  • Very good thermal resistance
  • Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
The flat flange bell is used for tests with a vacuum experimental plate (vacuum chamber). The bell has a ground flange and a ground joint for the insertion of measuring equipment.