Evaporating dishes with snap cap
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1677 1678
KART1677EA 143 EUR
KART1677 KART1678
Evaporating dishes with snap cap
Reservoirs/Schalen/Potten Uitdampschalen
Made of PFA, translucent.
  • Very good chemical resistance
  • Very good temperature resistance